Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: skpt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171121.5 | NP_741136.1 | 5 - 6 | 1038 | 71 | |
NM_171865.6 | NP_741137.1 | 5 - 6 | 1071 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |