Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: skpt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171121.5 | NP_741136.1 | 6 - 7 | 1205 | 78 | |
NM_171865.6 | NP_741137.1 | 6 - 7 | 1238 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |