Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mdt-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001382882.1 | 4 - 5 | 1004 | 95 | ||
NM_171134.8 | NP_741151.1 | 4 - 5 | 1033 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |