Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mdt-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001382882.1 | 6 - 7 | 2473 | 76 | ||
NM_171134.8 | NP_741151.1 | 6 - 7 | 2502 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |