Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cct-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171135.9 | NP_741153.1 | 3 - 4 | 341 | 82 | |
NM_171136.6 | NP_741154.1 | 3 - 4 | 336 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |