Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lim-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066082.5 | NP_498483.1 | 13 - 14 | 2586 | 90 | |
NM_066083.7 | NP_498484.1 | 12 - 13 | 2276 | 90 | ||
NM_171173.4 | NP_741201.1 | 16 - 17 | 2814 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |