Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aco-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066337.7 | NP_498738.2 | 3 - 4 | 435 | 70 | |
NM_171200.5 | NP_741235.1 | 3 - 4 | 432 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |