Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gei-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066372.7 | NP_498773.3 | 17 - 18 | 5356 | 68 | |
NM_171207.8 | NP_741245.3 | 16 - 17 | 5107 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |