Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hipr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066524.6 | NP_498925.1 | 7 - 8 | 1708 | 108 | |
NM_171215.7 | NP_741253.1 | 7 - 8 | 1709 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |