Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027810.5 | NP_001022981.1 | 2 - 3 | 258 | 93 | |
NM_001129276.3 | NP_001122748.1 | 2 - 3 | 258 | 93 | ||
NM_001268079.4 | NP_001255008.1 | 2 - 3 | 258 | 93 | ||
NM_001306680.3 | NP_001293609.1 | 2 - 3 | 194 | 93 | ||
NM_001306682.3 | NP_001293611.1 | 1 - 2 | 86 | 93 | ||
NM_001306684.3 | NP_001293613.1 | 2 - 3 | 194 | 93 | ||
NM_001306685.3 | NP_001293614.1 | 2 - 3 | 194 | 93 | ||
NM_001306686.3 | NP_001293615.1 | 2 - 3 | 194 | 93 | ||
NM_001382965.1 | 2 - 3 | 258 | 93 | |||
NM_171221.7 | NP_741263.3 | 2 - 3 | 295 | 93 | ||
GenBank mRNA | AF424978.1 | - | 2 - 3 | 194 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |