Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027810.5 | NP_001022981.1 | 3 - 4 | 540 | 70 | |
NM_001129276.3 | NP_001122748.1 | 3 - 4 | 540 | 70 | ||
NM_001268079.4 | NP_001255008.1 | 3 - 4 | 540 | 70 | ||
NM_001306680.3 | NP_001293609.1 | 3 - 4 | 476 | 70 | ||
NM_001306682.3 | NP_001293611.1 | 2 - 3 | 368 | 70 | ||
NM_001306684.3 | NP_001293613.1 | 3 - 4 | 476 | 70 | ||
NM_001306685.3 | NP_001293614.1 | 3 - 4 | 476 | 70 | ||
NM_001306686.3 | NP_001293615.1 | 3 - 4 | 476 | 70 | ||
NM_001382965.1 | 3 - 4 | 540 | 70 | |||
NM_171221.7 | NP_741263.3 | 3 - 4 | 577 | 70 | ||
GenBank mRNA | AF424978.1 | - | 3 - 4 | 476 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |