Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cor-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171226.5 | NP_741268.1 | 7 - 8 | 1509 | 61 | |
NM_171892.7 | NP_741269.1 | 7 - 8 | 1490 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |