Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tra-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027709.3 | NP_001022880.2 | 17 - 18 | 3322 | 79 | |
GenBank mRNA | M93256.1 | - | 17 - 18 | 3337 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |