Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eri-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171244.5 | NP_741292.1 | 2 - 3 | 465 | 76 | |
NM_171245.5 | NP_741293.1 | 2 - 3 | 465 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |