Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: inx-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171246.5 | NP_741294.1 | 1 - 2 | 156 | 66 | |
NM_171247.6 | NP_741295.1 | 1 - 2 | 138 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |