Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: inx-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171246.5 | NP_741294.1 | 3 - 4 | 349 | 104 | |
NM_171247.6 | NP_741295.1 | 3 - 4 | 331 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |