Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snn-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380065.1 | NP_001367952.1 | 2 - 3 | 207 | 65 | |
NM_001380066.2 | NP_001367951.1 | 2 - 3 | 226 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Caenorhabditis elegans Probe spans exons |