Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028052.5 | NP_001023223.1 | 9 - 10 | 2352 | 107 | |
NM_001028053.5 | NP_001023224.1 | 4 - 5 | 1444 | 107 | ||
NM_001307521.4 | NP_001294450.1 | 9 - 10 | 2255 | 107 | ||
NM_001380068.1 | NP_001368271.1 | 4 - 5 | 1358 | 107 | ||
NM_067740.6 | NP_500141.1 | 9 - 10 | 2246 | 107 | ||
NM_067741.5 | NP_500142.1 | 9 - 10 | 2117 | 107 | ||
NM_171279.6 | NP_741329.1 | 9 - 10 | 2153 | 107 |
Gene Symbol: ccm-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380067.2 | NP_001367894.1 | 18 - 19 | 3144 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Caenorhabditis elegans Probe spans exons |