Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ser-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307333.2 | NP_001294262.1 | 3 - 4 | 679 | 77 | |
NM_171297.4 | NP_741350.1 | 3 - 4 | 654 | 77 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |