Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: soc-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026088.4 | NP_001021259.1 | 5 - 6 | 981 | 112 | |
NM_001306829.3 | NP_001293758.1 | 6 - 7 | 1134 | 112 | ||
NM_001306830.3 | NP_001293759.1 | 6 - 7 | 925 | 112 | ||
NM_171332.5 | NP_741391.2 | 6 - 7 | 899 | 112 | ||
GenBank mRNA | AF054827.1 | - | 5 - 6 | 988 | 112 | |
AF068919.1 | - | 5 - 6 | 894 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |