Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mgl-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083266.5 | NP_001076735.3 | 8 - 9 | 1709 | 123 | |
NM_001313057.3 | NP_001299986.1 | 2 - 3 | 409 | 123 | ||
NM_001313058.3 | NP_001299987.1 | 2 - 3 | 409 | 123 | ||
NM_171341.5 | NP_741400.2 | 8 - 9 | 1707 | 123 | ||
NR_132462.1 | 8 - 9 | 1639 | 123 | |||
NR_132463.1 | 8 - 9 | 1639 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |