Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lag-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028107.5 | NP_001023278.1 | 10 - 11 | 1415 | 80 | |
NM_001306811.4 | NP_001293740.1 | 8 - 9 | 3404 | 80 | ||
NM_001380259.1 | NP_001368385.1 | 8 - 9 | 1274 | 80 | ||
NM_171350.7 | NP_741410.1 | 10 - 11 | 1421 | 80 | ||
GenBank mRNA | U49794.1 | - | 10 - 11 | 1419 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |