Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: csp-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380264.1 | NP_001368632.1 | 8 - 9 | 1611 | 85 | |
NM_001380265.1 | NP_001368631.1 | 1 - 2 | 219 | 85 | ||
NM_001383103.1 | 11 - 12 | 1913 | 85 | |||
GenBank mRNA | AF088288.1 | - | 11 - 12 | 1910 | 85 | |
AF088289.1 | - | 1 - 2 | 219 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |