Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: egl-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027908.6 | NP_001023079.1 | 8 - 9 | 3890 | 63 | |
NM_171379.6 | NP_741442.1 | 8 - 9 | 3890 | 63 | ||
GenBank mRNA | AF023602.1 | - | 9 - 10 | 3923 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |