Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-94 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383183.1 | 5 - 6 | 492 | 112 | ||
NM_059334.5 | NP_491735.1 | 3 - 4 | 375 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |