Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: csn-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059339.9 | NP_491740.1 | 1 - 2 | 58 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |