Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: csn-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059339.9 | NP_491740.1 | 5 - 6 | 1318 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |