Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vps-34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380450.1 | NP_001367304.1 | 3 - 4 | 1446 | 73 | |
NM_001383194.1 | 2 - 3 | 1661 | 73 | |||
NM_059340.5 | NP_491741.1 | 3 - 4 | 1458 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |