Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ceh-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059345.6 | NP_491746.1 | 2 - 3 | 407 | 109 | |
GenBank mRNA | AY246428.1 | - | 2 - 3 | 382 | 109 | |
AY246429.1 | - | 2 - 3 | 360 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |