Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: haf-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059353.7 | NP_491754.1 | 1 - 2 | 128 | 72 | |
NM_182010.5 | NP_871810.1 | 1 - 2 | 111 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |