Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: haf-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059353.7 | NP_491754.1 | 5 - 6 | 786 | 87 | |
NM_182010.5 | NP_871810.1 | 5 - 6 | 769 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |