Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cof-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059403.5 | NP_491804.2 | 2 - 3 | 571 | 65 | |
GenBank mRNA | AY494975.1 | - | 2 - 3 | 475 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |