Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ima-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059423.7 | NP_491824.1 | 5 - 6 | 1441 | 140 | |
GenBank mRNA | AF040996.1 | - | 4 - 5 | 1399 | 140 | |
AF326936.1 | - | 4 - 5 | 1403 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |