Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mut-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001372960.2 | NP_001360009.1 | 5 - 6 | 820 | 112 | |
NM_059433.8 | NP_491834.1 | 5 - 6 | 819 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |