Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snt-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059452.8 | NP_491853.2 | 3 - 4 | 405 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |