Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: che-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083128.1 | NP_001076597.1 | 1 - 2 | 70 | 97 | |
NM_001083129.2 | NP_001076598.1 | 1 - 2 | 109 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Caenorhabditis elegans Probe spans exons |