Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acdh-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313506.3 | NP_001300435.1 | 2 - 3 | 300 | 66 | |
NM_059485.5 | NP_491886.2 | 1 - 2 | 58 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |