Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mei-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059493.7 | NP_491894.1 | 2 - 3 | 419 | 124 | |
GenBank mRNA | AF248052.1 | - | 2 - 3 | 401 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |