Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ckr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059517.4 | NP_491918.3 | 2 - 3 | 295 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 G-protein coupled receptor Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |