Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059531.8 | NP_491932.1 | 2 - 3 | 235 | 74 | |
GenBank mRNA | AF001271.1 | - | 2 - 3 | 245 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |