Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpc-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059534.8 | NP_491935.1 | 1 - 2 | 105 | 64 | |
GenBank mRNA | AF291849.1 | - | 1 - 2 | 93 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |