Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpf-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059555.2 | NP_491956.1 | 16 - 17 | 2254 | 73 | |
NM_059556.5 | NP_491957.1 | 16 - 17 | 2242 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |