Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059617.6 | NP_492018.1 | 5 - 6 | 1397 | 70 | |
GenBank mRNA | AB000913.1 | - | 5 - 6 | 1415 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |