Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059617.6 | NP_492018.1 | 7 - 8 | 1720 | 125 | |
GenBank mRNA | AB000913.1 | - | 7 - 8 | 1738 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |