Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306537.3 | NP_001293466.1 | 7 - 8 | 1304 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |