Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eat-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059667.7 | NP_492068.1 | 7 - 8 | 1280 | 143 | |
GenBank mRNA | U59210.1 | - | 7 - 8 | 1257 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |