Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059684.6 | NP_492085.1 | 3 - 4 | 370 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |