Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pbrm-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025837.4 | NP_001021008.1 | 13 - 14 | 5402 | 115 | |
NM_001383360.1 | 13 - 14 | 5402 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |