Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pbrm-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025837.4 | NP_001021008.1 | 5 - 6 | 2947 | 82 | |
NM_001383360.1 | 5 - 6 | 2955 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |