Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glb-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001136283.3 | NP_001129755.1 | 6 - 7 | 1074 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |